Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzylacetone 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17647-0500 17647-2500
TS17647-0500EA 93 USD InStock
TS17647-0500 TS17647-2500
4-Hydroxybenzylacetone 99+%
4-Hydroxybenzylacetone


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 292
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00002394
CAS Number: 5471-51-2
EINECS: 226-806-4
Order Now
Specification Test Results
Appearance White to light yellow Powder or needles
Infrared spectrum Conforms
Melting point 81 to 85 °C
GC ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty