Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromobenzyl alcohol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18010-0100 18010-0250
TS18010-0100EA 46 USD InStock
TS18010-0100 TS18010-0250
2-Bromobenzyl alcohol 98%
2-Bromobenzyl alcohol


Formula: C₇H₇BrO
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 78…82 °C
MDL Number: MFCD00004600
CAS Number: 18982-54-2
EINECS: 242-719-4
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Melting point 78 to 82 °C
GC ≥97.5 %
HPLC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty