Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Hydroxyphenyl)ethanol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

18249-0010 18249-0050
TS18249-0010EA 38 USD InStock
TS18249-0010 TS18249-0050
2-(2-Hydroxyphenyl)ethanol 98%
2-(2-Hydroxyphenyl)ethanol


Formula: C₈H₁₀O₂
Boiling Pt: 168…169 °C (2 mmHg)
Density: 1.159 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002890
CAS Number: 7768-28-7
Order Now
Specification Test Results
Appearance Clear yellow to yellow-brown Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5560 to 1.5600 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥96.0 %
Specific optical rotation -125° to -142° (20°C, 589 nm) (0.25 % in Methanol)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty