Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4-Trimethoxybenzoic acid 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18347-0250 18347-1000
TS18347-0250EA 34 USD InStock
TS18347-0250 TS18347-1000
2,3,4-Trimethoxybenzoic acid 98+%
2,3,4-Trimethoxybenzoic acid


Formula: C₁₀H₁₂O₅
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002433
CAS Number: 573-11-5
EINECS: 209-350-0
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 102 °C
Titration with NaOH ≥98.0 %
Residue after ignition ≤0.2 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty