Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18671-0010 18671-0050 18671-2500
TS18671-0010EA 154 USD InStock
TS18671-0010 TS18671-0050 TS18671-2500
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 98%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride


Formula: C₇H₄F₅NO
Melting Pt: 227 °C (subl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012953
CAS Number: 57981-02-9
Order Now
Specification Test Results
Appearance White Crystalline powder, crystals, flakes and/or chunks
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Composition ≤2 % (CoCl2) Typical

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty