Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Diphenylethanol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18822-0020 18822-0100
TS18822-0020EA 36 USD InStock
TS18822-0020 TS18822-0100
2,2-Diphenylethanol 97%
2,2-Diphenylethanol


Formula: C₁₄H₁₄O
Boiling Pt: 160…162 °C (5 mmHg)
Melting Pt: 53…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004729
CAS Number: 1883-32-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 61 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty