Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-2-fluorotoulene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

19796-0050 19796-0250
TS19796-0050EA 35 USD InStock
TS19796-0050 TS19796-0250
α-Bromo-2-fluorotoulene 98%
α-Bromo-2-fluorotoulene


Formula: C₇H₆BrF
Boiling Pt: 84…85 °C (15 mmHg)
Density: 1.567 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000324
CAS Number: 446-48-0
EINECS: 207-169-1
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5495 to 1.5515 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty