Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-4-fluorobenzene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20820-0050 20820-0250
TS20820-0050EA 20 USD InStock
TS20820-0050 TS20820-0250
1-Chloro-4-fluorobenzene 98%
1-Chloro-4-fluorobenzene


Formula: C₆H₄ClF
MW: 130.55
Boiling Pt: 129
Melting Pt: -21.5
Density: 1.226 g/cm³
Flash Pt: 29 °C
MDL Number: MFCD00000603
CAS Number: 352-33-0
EINECS: 206-521-1
UN: 1993
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4960 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty