Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-3-Bromo-1,2-propanediol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

20985-0050 20985-0250
TS20985-0050EA 101 USD InStock
TS20985-0050 TS20985-0250
(±)-3-Bromo-1,2-propanediol 97%
(±)-3-Bromo-1,2-propanediol


Formula: BrH₂CCH(OH)CH₂OH
Boiling Pt: 72…75 °C (.2 mmHg)
Density: 1.77 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004711
CAS Number: 4704-77-2
EINECS: 225-186-2
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5165 to 1.5195 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty