Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorobenzonitrile 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

29049-0050 29049-0010 29049-0250
TS29049-0050EA 70 USD InStock
TS29049-0050 TS29049-0010 TS29049-0250
4-Bromo-2-fluorobenzonitrile 99+%
4-Bromo-2-fluorobenzonitrile


Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 238…239 °C (1013 hPa)
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00143264
CAS Number: 105942-08-3
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to light brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 69 to 72 °C
GC ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty