Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Vinylbenzoic acid 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

29491-0010 29491-0050
TS29491-0010EA 74 USD InStock
TS29491-0010 TS29491-0050
4-Vinylbenzoic acid 96%
4-Vinylbenzoic acid


Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 282
Melting Pt: 115
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
EINECS: 214-053-4
Order Now
Specification Test Results
Appearance Cream to brown Powder
Infrared spectrum Conforms
Solubility (10% in Methanol) Clear solution
Titration with NaOH ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty