Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzhydrylamine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30432-0250 30432-1000
TS30432-0250EA 76 USD InStock
TS30432-0250 TS30432-1000
Benzhydrylamine 97%
Benzhydrylamine


Formula: C₁₃H₁₃N
Boiling Pt: 295 °C (760 mmHg)
Melting Pt: 12 °C
Density: 1.06 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008059
CAS Number: 91-00-9
EINECS: 202-032-2
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5950 to 1.5970 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty