Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoro-4-iodotoluene 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30618-0050 30618-0010
TS30618-0050EA 81 USD InStock
TS30618-0050 TS30618-0010
α,α,α-Trifluoro-4-iodotoluene 97%
α,α,α-Trifluoro-4-iodotoluene


Formula: C₇H₄F₃I
Boiling Pt: 76…79 °C (15 mmHg)
Melting Pt: 17 °C
Density: 1.851 g/cm³
Flash Pt: 68 °C
MDL Number: MFCD00039398
CAS Number: 455-13-0
EINECS: 207-234-4
Order Now
Specification Test Results
Appearance Clear colorless to yellow to red Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5200 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty