Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromophenyl isocyanate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

31413-0050 31413-0010
TS31413-0050EA 87 USD InStock
TS31413-0050 TS31413-0010
3-Bromophenyl isocyanate 97%
3-Bromophenyl isocyanate


Formula: C₇H₄BrNO
Boiling Pt: 220 °C (760 mmHg)
Density: 1.586 g/cm³
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002014
CAS Number: 23138-55-8
UN: 2206
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5835 to 1.5865 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty