Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)phenyl Isocyanate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

31417-0050 31417-0010
TS31417-0050EA 120 USD InStock
TS31417-0050 TS31417-0010
3,5-Bis(trifluoromethyl)phenyl Isocyanate 98%
3,5-Bis(trifluoromethyl)phenyl Isocyanate


Formula: C₉H₃F₆NO
Boiling Pt: 62 °C (2 mmHg)
Density: 1.47 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00013559
CAS Number: 16588-74-2
UN: 2206
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4295 to 1.4315 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty