Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-3-Amino-1,2-propanediol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

39788-0050 39788-0250
TS39788-0050EA 134 USD InStock
TS39788-0050 TS39788-0250
(R)-(+)-3-Amino-1,2-propanediol 98%
(R)-(+)-3-Amino-1,2-propanediol


Formula: C₃H₉NO₂
Boiling Pt: 163 °C (15 mmHg)
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00798261
CAS Number: 66211-46-9
UN: 3259
ADR: 8,II
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
Specific optical rotation +27° to +29° (20 °C, 589 nm) (C=4, 5 N HCl)
Titration with HClO4 ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty