Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2S)-(-)-2-Amino-1,2-diphenylethanol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39789-0010 39789-0050
TS39789-0010EA 67 USD InStock
TS39789-0010 TS39789-0050
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol 99%
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol


Formula: C₁₄H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD00074960
CAS Number: 23190-16-1
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 141 to 145 °C
HPLC ≥98.5 %
Specific optical rotation -6° to -9° (20 °C, 589 nm) (C=0.5%, Ethanol)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty