Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Vinylbenzyl acetate 95% stabilized

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42250-0050 42250-0250
TS42250-0050EA 87 USD InStock
TS42250-0050 TS42250-0250
4-Vinylbenzyl acetate 95% stabilized
4-Vinylbenzyl acetate


Formula: C₁₁H₁₂O₂
Boiling Pt: 125 °C (12 mmHg)
Density: 1.53 g/cm³
Flash Pt: >93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078265
CAS Number: 1592-12-7
Order Now
Specification Test Results
Appearance Clear light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.5355 to 1.5385 (20°C, 589 nm)
GC ≥94 %
Stabilizer 0.1 % hydroquinone

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty