Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Diaminophenol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42568-0010 42568-0050
TS42568-0010EA 89 USD InStock
TS42568-0010 TS42568-0050
2,3-Diaminophenol 97%
2,3-Diaminophenol


Formula: C₆H₈N₂O
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00075199
CAS Number: 59649-56-8
Order Now
Specification Test Results
Appearance Light brown to dark brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 161 to 165 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty