Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 90%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

42690-0050 42690-0010
TS42690-0050EA 237 USD InStock
TS42690-0050 TS42690-0010
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 90%
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene


Formula: C₁₃H₂₀N₂O₂
MW: 236.3100 g/mol
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD02683058
CAS Number: 108468-00-4
UN: 3263
ADR: 8 III
Order Now
Specification Test Results
Appearance White to brown Crystalline powder or solid
Infrared spectrum Conforms
GC ≥89.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty