Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-3-hydroxy-4-pyrone 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45080-1000 45080-0250
TS45080-1000EA 82 USD InStock
TS45080-1000 TS45080-0250
2-Ethyl-3-hydroxy-4-pyrone 98%
2-Ethyl-3-hydroxy-4-pyrone


Formula: C₇H₈O₃
Boiling Pt: 289…290 °C
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00059795
CAS Number: 4940-11-8
Order Now
Specification Test Results
Appearance White to cream Crystalline powder or solid
Infrared spectrum Conforms
Melting point 88 to 94 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty