Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-6-methylbenzoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45306-0010 45306-0050 45306-0250
TS45306-0010EA 70 USD InStock
TS45306-0010 TS45306-0050 TS45306-0250
2-Hydroxy-6-methylbenzoic acid 98%
2-Hydroxy-6-methylbenzoic acid


Formula: C₈H₈O₃
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD01194284
CAS Number: 567-61-3
Order Now
Specification Test Results
Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
Melting point 168 to 176 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty