Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-methylpyrimidine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10409-0100 10409-1000
TS10409-0100EA 26 USD InStock
TS10409-0100 TS10409-1000
2-Amino-4-methylpyrimidine 97%
2-Amino-4-methylpyrimidine


Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 283
Melting Pt: 155…160 °C
MDL Number: MFCD00006101
CAS Number: 108-52-1
EINECS: 203-591-5
Order Now
Specification Test Results
Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty