Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-nitropyridine 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10423-0050 10423-0250 10423-1000 10423-2500
TS10423-0050EA 34 USD InStock
TS10423-0050 TS10423-0250 TS10423-1000 TS10423-2500
2-Amino-5-nitropyridine 99%
2-Amino-5-nitropyridine


Formula: C₅H₅N₃O₂
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00006325
CAS Number: 4214-76-0
EINECS: 224-145-6
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 186 to 190 °C
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty