Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,1,3-Benzothiadiazole 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10566-0050 10566-0250 10566-1000
TS10566-0050EA 41 USD InStock
TS10566-0050 TS10566-0250 TS10566-1000
2,1,3-Benzothiadiazole 98%
2,1,3-Benzothiadiazole


Formula: C₆H₄N₂S
Boiling Pt: 206 °C (760 mmHg)
Melting Pt: 42…44 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00005809
CAS Number: 273-13-2
Order Now
Specification Test Results
Appearance Colorless to white to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty