Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cloxiquina 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10930-0050 10930-1000
TS10930-0050EA 22 USD InStock
TS10930-0050 TS10930-1000
Cloxiquina 95%
Cloxiquina


Formula: C₉H₆ClNO
MW: 179.61 g/mol
Boiling Pt: 349
Melting Pt: 125
Storage Temperature: Ambient
MDL Number: MFCD00006788
CAS Number: 130-16-5
EINECS: 204-978-1
Merck Index: 12,02483
Order Now
Specification Test Results
Appearance Light green to grey Powder
Infrared spectrum Conforms
Melting point 121 to 126 °C
Titration with HClO4 ≥94 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty