Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloropyridine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11373-0050 11373-1000
TS11373-0050EA 33 USD InStock
TS11373-0050 TS11373-1000
2,6-Dichloropyridine 98%
2,6-Dichloropyridine


Formula: C₅H₃Cl₂N
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 84…89 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006244
CAS Number: 2402-78-0
EINECS: 219-282-3
Order Now
Specification Test Results
Appearance White to pink to light grey Crystalline powder
Infrared spectrum Conforms
Melting point 84 to 89 °C
GC ≥97.5 %
Iodate (IO3) ≤3 ppm
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Barium (Ba) ≤0.002 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.01 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty