Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diglycolic anhydride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11467-0250 11467-1000
TS11467-0250EA 129 USD InStock
TS11467-0250 TS11467-1000
Diglycolic anhydride 97%
Diglycolic anhydride


Formula: C₄H₄O₄
MW: 116.07 g/mol
Melting Pt: 90…95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006677
CAS Number: 4480-83-5
EINECS: 224-761-5
Order Now
Specification Test Results
Appearance White to cream Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 90 to 95 °C
Titration excess NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty