Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethyl-3-ethylpyrrole 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11621-0050 11621-0250
TS11621-0050EA 131 USD InStock
TS11621-0050 TS11621-0250
2,4-Dimethyl-3-ethylpyrrole 96%
2,4-Dimethyl-3-ethylpyrrole


Formula: C₈H₁₃N
Boiling Pt: 197 °C (710 mmHg)
Density: 0.913 g/cm³
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005222
CAS Number: 517-22-6
EINECS: 208-234-7
Order Now
Specification Test Results
Appearance Clear yellow to red-brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4980 (20°C, 589 nm)
GC ≥95 %
Titration excess NaOH ≥97.0 %
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty