Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethylmaleic anhydride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11639-0050 11639-0500
TS11639-0050EA 92 USD InStock
TS11639-0050 TS11639-0500
2,3-Dimethylmaleic anhydride 97%
2,3-Dimethylmaleic anhydride


Formula: C₆H₆O₃
Melting Pt: 91…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00005523
CAS Number: 766-39-2
EINECS: 212-165-8
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 91 to 96 °C
Titration with TBAH ≥96.0 %
Chloride (Cl) ≤5 ppm
Nitrite (NO2) (Limit about 5 ppm) Passes test
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty