Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-1H-pyrazole 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11670-1000 11670-5000
TS11670-1000EA 72 USD InStock
TS11670-1000 TS11670-5000
3,5-Dimethyl-1H-pyrazole 99%
3,5-Dimethyl-1H-pyrazole


Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 218
Melting Pt: 106…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00005243
CAS Number: 67-51-6
EINECS: 200-657-5
Order Now
Specification Test Results
Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 106 to 109 °C
Titration with HClO4 ≥98.5 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.01 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 % (in methanol)
Non-volatiles with MeOH ≤0.05 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty