Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxyindole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12127-0010 12127-0050 12127-2500
TS12127-0010EA 41 USD InStock
TS12127-0010 TS12127-0050 TS12127-2500
5-Hydroxyindole 97%
5-Hydroxyindole


Formula: C₈H₇NO
Melting Pt: 105…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005677
CAS Number: 1953-54-4
EINECS: 217-782-6
Order Now
Specification Test Results
Appearance Beige to brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 105 to 110 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty