Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12215-0025 12215-0100 12215-0250 12215-1000 12215-5000
TS12215-0025EA 844 USD InStock
TS12215-0025 TS12215-0100 TS12215-0250 TS12215-1000 TS12215-5000
Indole 99+%
Indole


Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253…254 °C (760 mmHg)
Melting Pt: 51…53 °C
Density: 1.22 g/cm³
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,04985
Order Now
Specification Test Results
Appearance White to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 53 °C
GC ≥99.0 %
UV at 195 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty