Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl nicotinate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12721-1000 12721-5000
TS12721-1000EA 33 USD InStock
TS12721-1000 TS12721-5000
Methyl nicotinate 99%
Methyl nicotinate


Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 209
Melting Pt: 39
Flash Pt: 95 °C
MDL Number: MFCD00006388
CAS Number: 93-60-7
EINECS: 202-261-8
Merck Index: 12,06174
Order Now
Specification Test Results
Appearance White to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 38 to 43 °C
Titration with HClO4 ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty