Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-3-pyrrolidinol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12762-0010 12762-0050
TS12762-0010EA 104 USD InStock
TS12762-0010 TS12762-0050
N-Methyl-3-pyrrolidinol 97%
N-Methyl-3-pyrrolidinol


Formula: C₅H₁₁NO
Boiling Pt: 50…52 °C (1 mmHg)
Density: 0.921 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00003176
CAS Number: 13220-33-2
Order Now
Specification Test Results
Appearance Clear colorless to yellow to golden Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4650 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty