Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazine-2-carboxylic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

13171-0250 13171-1000
TS13171-0250EA 43 USD InStock
TS13171-0250 TS13171-1000
Pyrazine-2-carboxylic acid 99%
Pyrazine-2-carboxylic acid


Formula: C₅H₄N₂O₂
Boiling Pt: 313 °C (1013 hPa)
MDL Number: MFCD00006130
CAS Number: 98-97-5
EINECS: 202-718-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty