Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazole 98%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

13174-0250 13174-1000
TS13174-0250EA 66 USD InStock
TS13174-0250 TS13174-1000
1H-Pyrazole 98%, pure
1H-Pyrazole


Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00005234
CAS Number: 288-13-1
EINECS: 206-017-1
Merck Index: 12,08144
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 66 to 70 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty