Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-2,4-dicarboxylic acid hydrate 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

13186-0010 13186-0050 13186-0250
TS13186-0010EA 24.72 USD InStock
TS13186-0010 TS13186-0050 TS13186-0250
Pyridine-2,4-dicarboxylic acid hydrate 99+%
Pyridine-2,4-dicarboxylic acid hydrate


Formula: C₇H₅NO₄·xH₂O
Melting Pt: 243…246 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149413
CAS Number: 499-80-9
EINECS: 207-892-2
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤10 % (K.F.)
Titration with NaOH ≥99.0 % (On dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty