Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methylpyridin-3-ol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

14811-0250 14811-1000
TS14811-0250EA 76 USD InStock
TS14811-0250 TS14811-1000
6-Methylpyridin-3-ol 99%
6-Methylpyridin-3-ol


Formula: C₆H₇NO
Melting Pt: 168…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00006339
CAS Number: 1121-78-4
EINECS: 214-337-8
Order Now
Specification Test Results
Appearance Beige to tan Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 171 °C
Titration with HClO4 ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty