Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromopyridine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15374-0050 15374-0250
TS15374-0050EA 34 USD InStock
TS15374-0050 TS15374-0250
2-Amino-5-bromopyridine 97%
2-Amino-5-bromopyridine


Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 244
Melting Pt: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00006323
CAS Number: 1072-97-5
EINECS: 214-019-9
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 135 to 140 °C
GC ≥96.0 %
Residue after evaporation ≤0.002 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty