Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Citraconic anhydride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15479-0010 15479-0250 15479-1000 15479-2500
TS15479-0010EA 477 USD InStock
TS15479-0010 TS15479-0250 TS15479-1000 TS15479-2500
Citraconic anhydride 98%
Citraconic anhydride


Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 213
Melting Pt: 6…7 °C
Density: 1.245 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00005522
CAS Number: 616-02-4
EINECS: 210-459-0
UN: 2922
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4690 to 1.4730 (20°C, 589 nm)
Titration with NaOH 97.5 to 102.5 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty