Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-nitrothiazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15852-0250 15852-1000
TS15852-0250EA 36 USD InStock
TS15852-0250 TS15852-1000
2-Amino-5-nitrothiazole 97%
2-Amino-5-nitrothiazole


Formula: C₃H₃N₃O₂S
Melting Pt: 196…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00005326
CAS Number: 121-66-4
EINECS: 204-490-9
Merck Index: 13,00456
Order Now
Specification Test Results
Appearance Yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Identification Passes test
Relative density 0.790 to 0.793
Residue after evaporation ≤50 ppm
Related substances ≤2 ppm (Benzene)
Acidity/alkalinity Passes test
Insoluble matter (in water) Passes test
Reducing substances Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty