Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Primaquine diphosphate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17087-0010 17087-0050
TS17087-0010EA 52 USD InStock
TS17087-0010 TS17087-0050
Primaquine diphosphate 98%
Primaquine diphosphate


Formula: C₁₅H₂₇N₃O₉P₂
Melting Pt: 205…206 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013166
CAS Number: 63-45-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Orange to red-orange Powder
Infrared spectrum Conforms
Melting point 199 to 205 °C
Titration with NaOH 97.5 to 102.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty