Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloro-3-nitropyridine 90% tech.

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18600-0100 18600-0500 18600-2500
TS18600-0100EA 30 USD InStock
TS18600-0100 TS18600-0500 TS18600-2500
2,6-Dichloro-3-nitropyridine 90% tech.
2,6-Dichloro-3-nitropyridine


Formula: C₅H₂Cl₂N₂O₂
Melting Pt: 58…68 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006234
CAS Number: 16013-85-7
EINECS: 240-151-1
Order Now
Specification Test Results
Appearance Yellow to orange-brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 58 to 68 °C
Water ≤10 % (K.F.)
GC ≥88 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty