Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloropyridine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18614-0050 18614-0250 18614-1000
TS18614-0050EA 42 USD InStock
TS18614-0050 TS18614-0250 TS18614-1000
2,5-Dichloropyridine 98%
2,5-Dichloropyridine


Formula: C₅H₃Cl₂N
Boiling Pt: 190…191 °C (760 mmHg)
Melting Pt: 58…62 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006239
CAS Number: 16110-09-1
EINECS: 240-278-2
Order Now
Specification Test Results
Appearance White to light yellow Crystalline solid
Infrared spectrum Conforms
Melting point 58 to 62 °C
Water ≤1 %
GC ≥97.5 %
Titration with NaOH 28.1 to 29.5 % (citric acid dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty