Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydrofuran 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

19044-1000 19044-5000
TS19044-1000EA 75 USD InStock
TS19044-1000 TS19044-5000
2,3-Dihydrofuran 98+%
2,3-Dihydrofuran


Formula: C₄H₆O
Density: 0.927 g/cm³
Flash Pt: –24 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003205
CAS Number: 1191-99-7
EINECS: 214-747-7
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4250 (20°C, 589 nm)
Water ≤0.3 %
GC ≥98.0 %
Tetrahydrofuran ≤1.0 % (GC)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty