Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylbenzofuran 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20840-0010 20840-0050
TS20840-0010EA 118 USD InStock
TS20840-0010 TS20840-0050
2-Methylbenzofuran 96%
2-Methylbenzofuran


Formula: C₉H₈O
Boiling Pt: 197…198 °C (760 mmHg)
Density: 1.057 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00005850
CAS Number: 4265-25-2
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty