Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylimidazole 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21928-0025 21928-1000 21928-5000
TS21928-0025EA 280 USD InStock
TS21928-0025 TS21928-1000 TS21928-5000
2-Ethylimidazole 99%
2-Ethylimidazole


Formula: C₅H₈N₂
Boiling Pt: 268 °C (760 mmHg)
Melting Pt: 83…87 °C
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00005192
CAS Number: 1072-62-4
EINECS: 214-011-5
Order Now
Specification Test Results
Appearance White to light yellow Flakes
Infrared spectrum Conforms
Melting point 83 to 87 °C
Water ≤0.5 %
GC ≤1.0 % Imidazole

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty