Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-Epoxystyrene 95%, 95% ee

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

24055-0010 24055-0050 24055-0250
TS24055-0010EA 47 USD InStock
TS24055-0010 TS24055-0050 TS24055-0250
(R)-(+)-Epoxystyrene 95%, 95% ee
(R)-(+)-Epoxystyrene


Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 192…195 °C (760 mmHg)
Melting Pt: –37 °C
Density: 1.053 g/cm³
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066210
CAS Number: 20780-53-4
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5380 (20°C, 589 nm)
GC ≥94 %
Enantiomeric excess ≥94 %
Specific optical rotation +31° to +35° (20°C, 589 nm) (neat)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty