Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-3-oxetanemethanol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

24084-0100 24084-0500
TS24084-0100EA 45 USD InStock
TS24084-0100 TS24084-0500
3-Methyl-3-oxetanemethanol 98%
3-Methyl-3-oxetanemethanol


Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 122 °C (70 mmHg)
Density: 1.034 g/cm³
Flash Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010273
CAS Number: 3143-02-0
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty